Omeprazole 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
J62860.03
J62860.06
J62860.03EA
1860
CZK
J62860.03
J62860.06
Omeprazole 98%
Omeprazole
Purity: 98%
CAS No.: 73590-58-6
Molecular Formula: C17H19N3O3S
Formula Weight: 345.42
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 158-159°
MDL No.: MFCD00083192
Applications: A proton pump inhibitor
Vzorec:
C₁₇H₁₉N₃O₃S MW: 345,42 g/mol Bod tání: 158…159 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00083192 CAS číslo: 73590-58-6 Merck Index: 14,6845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...