Tisk…
EDTA 0.5 M in aqueous solution pH 8.0
Dodavatel: Thermo Fisher Scientific
Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
J62786.AK J62786.AP
J62786.AKEA 2320 CZK
J62786.AK J62786.AP
EDTA 0.5 M in aqueous solution pH 8.0
EDTA
CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00003541
Vzorec: C₁₀H₁₆N₂O₈
MW: 292.25
Bod varu: 614
Bod tání: 220
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS číslo: 60-00-4
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM