Tisk…

Mefenamic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-(2,3-Xylidino)benzoic acid, 2-((2,3-Dimethylphenyl)amino)benzoic acid, 2-(2,3-Dimethylphenylamino)benzoic acid, N-(2,3-Dimethylphenyl)anthranilic acid

J62705.14 J62705.22
J62705.14EA 1580 CZK
J62705.14 J62705.22
Mefenamic acid 98%
Mefenamic acid

Purity: 98% CAS No.: 61-68-7 Molecular Formula: C15H15NO2 Formula Weight: 214.29 Physical Form: Powder Appearance: Off-white MDL No.: MFCD00051721


Applications: An non-steroidal anti-inflammatory drug with antiproliferative activity against human colon cancer cells

Vzorec: C₁₅H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00051721
CAS číslo: 61-68-7
EINECS: 200-513-1
Merck Index: 14,5798

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR