Tolmetin sodium salt dihydrate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-Methyl-5-[p-Toluoyl]Pyrrole-2 Acetic Acid sodium salt dihydrate
J62663.03
J62663.06
J62663.14
J62663.03EA
1740
CZK
J62663.03
J62663.06
J62663.14
Tolmetin sodium salt dihydrate ≥98%
Tolmetin sodium salt dihydrate
Purity: 98+%
CAS No.: 64490-92-2
Molecular Formula: C15H14NNaO3.2H2O
Formula Weight: 315.30 (279.27anhy)
Physical Form: Powder
Appearance: Off-White
Solubility: Soluble in water
MDL No.: MFCD00150761
Applications: An NSAID compound
Vzorec:
C₁₅H₁₈NNaO₅ MW: 315.301 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150761 CAS číslo: 64490-92-2 EINECS: 252-687-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...