Tylosin tartrate 94%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Pharmasin, [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt
Purity: 95%
CAS No.: 1405-54-5
Molecular Formula: C46H77NO17.C4H6O6
Formula Weight: 1066.20
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to cream
MDL No.: MFCD00070094
Applications: Macrolide antibiotic
Vzorec:
C₄₆H₇₇NO₁₇·C₄H₆O₆ MW: 1066,20 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00070094 CAS číslo: 1405-54-5 EINECS: 215-781-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...