Mineral oil, white (petroleum), high purity
Dodavatel: Thermo Scientific
|
Nebezpečí
|
J62592.AE
J62592.AP
J62592.AEEA
1630
CZK
J62592.AE
J62592.AP
Mineral oil, white (petroleum), high purity
Mineral oil, white (petroleum)
CAS No.: 8042-47-5
Physical Form: Oily liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00131611
Applications: Suitable for PCR applications
Bod vzplanutí:
>110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS číslo: 8042-47-5 EINECS: 232-455-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...