Imetit dihydrobromide 98%
Dodavatel: Thermo Scientific
|
Varování
|
J62346.MF
J62346.MC
J62346.MFEA
13040
CZK
J62346.MF
J62346.MC
Imetit dihydrobromide 98%
Imetit dihydrobromide
Purity: 98%
CAS No.: 32385-58-3
Molecular Formula: C6H10N4S.2HBr
Formula Weight: 332.06
Physical Form: Powder
Appearance: Off-white
Solubility: Freely soluble in water, practically insoluble in acetone
MDL No.: MFCD00153816
Applications: An extremely potent, high affinity agonist at H3 and H4 receptors
Vzorec:
C₆H₁₀N₄S·2HBr MW: 332,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153816 CAS číslo: 32385-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...