1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
ODQ
J62250.MA
J62250.MF
J62250.MAEA
3340
CZK
J62250.MA
J62250.MF
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥99%
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one
Purity: 99+%
CAS No.: 41443-28-1
Molecular Formula: C9H5N3O2
Formula Weight: 187.15
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to pale solid
Melting Point: 160-170°
MDL No.: MFCD00792620
Applications: Selective inhibitor of nitric oxide-sensitive guanylyl cyclase
Vzorec:
C₉H₅N₃O₂ Bod tání: 160…170 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00792620 CAS číslo: 41443-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...