Tisk…

Amlodipine besylate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylic acid 3-Ethyl 5-Methyl Ester Benzenesulphon, 3-Ethyl 5-Methyl 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate Benzenesulphonate, 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate benzenesulphonate

J62164.03 J62164.06 J62164.14
J62164.03EA 3470 CZK
J62164.03 J62164.06 J62164.14
Amlodipine besylate ≥98%
Amlodipine besylate

Purity: 98+% CAS No.: 111470-99-6 Molecular Formula: C20H25ClN2O5.C6H5SO3H Formula Weight: 567.05 Physical Form: Solid Appearance: Off-white Melting Point: 195-204° MDL No.: MFCD00887594


Applications: An L-type calcium channel antagonist with potent antioxidant activity

Vzorec: C₂₀H₂₅ClN₂O₅
MW: 567,05 g/mol
Bod tání: 195…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00887594
CAS číslo: 111470-99-6
Merck Index: 14,491

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR