Tisk…

Disodium-β-glycerophosphate 200 mM in solution

Dodavatel: Thermo Scientific

Synonyms: β-glycerophosphate

J62121.AD J62121.AE
J62121.ADEA 2050 CZK
J62121.AD J62121.AE
Disodium-β-glycerophosphate 200 mM in solution
Disodium-β-glycerophosphate solutions

CAS No.: 819-83-0 Molecular Formula: C3H7Na2O6P Formula Weight: 216.04 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00002135

Vzorec: C₃H₇Na₂O₆P
MW: 216,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002135
CAS číslo: 819-83-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR