Tisk…

Ofloxacin

Dodavatel: Thermo Fisher Scientific
Varování

J62080.03 J62080.06
J62080.03EA 1300 CZK
J62080.03 J62080.06
Ofloxacin
Ofloxacin

CAS No.: 82419-36-1 Molecular Formula: C18H20FN3O4 Formula Weight: 361.37 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Appearance: White to yellow Melting Point: 270-275° Solubility: Soluble in acetic acid or water. Slightly soluble in methanol MDL No.: MFCD00226105


Applications: Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity

Vzorec: C₁₈H₂₀FN₃O₄
Bod tání: 270…275 °C
Storage Temperature: Freezer
MDL Number: MFCD00226105
CAS číslo: 82419-36-1
Merck Index: 14,6771

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR