Tisk…

Glimepiride

Dodavatel: Thermo Scientific

Synonyms: 3-Ethyl-4-methyl-N-(4-(N-((rel-(1r,4r)-4-methylcyclohexyl)carbamoyl)sulphamoyl)phenethyl)-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide

J62032.MC J62032.03
J62032.MCEA 2850 CZK
J62032.MC J62032.03
Glimepiride
Glimepiride

CAS No.: 93479-97-1 Molecular Formula: C24H34N4O5S Formula Weight: 490.62 Physical Form: Powder Appearance: White Melting Point: 212-215° Solubility: Soluble in DMSO MDL No.: MFCD00878417


Applications: A sulfonylurea hypoglecemic agent and potent potassium channel blocker

Vzorec: C₂₄H₃₄N₄O₅S
MW: 490,62 g/mol
Bod tání: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00878417
CAS číslo: 93479-97-1
Merck Index: 14,4440

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR