Tisk…

Tamsulosin hydrochloride ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-[(R)-2-[2-(2-Ethoxyphenoxy)ethylamino]propyl]]-2-methoxybenzenesulphonamide Hydrochloride

J61999.MA J61999.MB J61999.MC
J61999.MAEA 4050 CZK
J61999.MA J61999.MB J61999.MC
Tamsulosin hydrochloride ≥98%
Tamsulosin hydrochloride

Purity: 98+% CAS No.: 106463-17-6 Molecular Formula: C20H28N2O5S.HCl Formula Weight: 444.97 Physical Form: Powder Appearance: White Melting Point: 228-230° MDL No.: MFCD00922997


Applications: An alpha-1 adrenoceptor antagonist

Vzorec: C₂₀H₂₈N₂O₅S
MW: 444,97 g/mol
Bod tání: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00922997
CAS číslo: 106463-17-6
Merck Index: 14,9049

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR