Tisk…

Phosphoenolpyruvic acid tris(cyclohexylammonium)salt ≥99%

Dodavatel: Thermo Scientific

Synonyms: PEP-3CHA

J61943.03 J61943.06 J61943.14
J61943.03EA 1470 CZK
J61943.03 J61943.06 J61943.14
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt ≥99%
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt

Purity: 99+% CAS No.: 35556-70-8 Molecular Formula: C3H5O6P.3C6H13N Formula Weight: 465.58 Physical Form: Powder Appearance: White Beilstein Reference: 3752336 MDL No.: MFCD00004258


Applications: Metabolic intermediate in many kinase reactions

Vzorec: C₃H₅O₆P·3C₆H₁₄N
MW: 465,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004258
CAS číslo: 35556-70-8
EINECS: 252-618-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR