Tisk…

Enoxacin, crystalline powder

Dodavatel: Thermo Scientific

Synonyms: 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic acid

J61912.03 J61912.06 J61912.14
J61912.03EA 1300 CZK
J61912.03 J61912.06 J61912.14
Enoxacin, crystalline powder
Enoxacin

A quinolone antibacteral agent which acts on DNA gyrase.


Enoxacin is an antibacterial agent used in the treatment of gastro enteritis including infectious diarrhea, respiratory tract infections, gonorrhea and urinary tract infections.


Applications: A quinolone antibacteral agent which acts on DNA gyrase


Upozornení: Store in cool place. Keep the container tightly closed in a dry and well-ventilated place.

Vzorec: C₁₅H₁₇FN₄O₃
MW: 320,32 g/mol
Bod tání: 220…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00133308
CAS číslo: 74011-58-8
Merck Index: 14,3587

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR