Tisk…

HEPES hemisodium salt

Dodavatel: Thermo Scientific

Synonyms: N-2-Hydroxyethylpiperazine-N'-2-ethanesulphonic acid hemisodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid hemisodium salt

J61830.22 J61830.30 J61830.36
J61830.22EA 2890 CZK
J61830.22 J61830.30 J61830.36
HEPES hemisodium salt
HEPES hemisodium salt

CAS No.: 103404-87-1 Molecular Formula: (C8H17N2O4S)2Na Formula Weight: 497.58 Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00036463


Applications: A zwitterionic buffer

Vzorec: C₈H₁₇.₅N₂O₄S·½Na
MW: 260,284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS číslo: 103404-87-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR