Prazosin hydrochloride
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(furan-2-yl)methanone hydrochloride, Prazosin HCl
J61712.MD
J61712.03
J61712.MDEA
2850
CZK
J61712.MD
J61712.03
Prazosin hydrochloride
Prazosin hydrochloride
CAS No.: 19237-84-4
Molecular Formula: C19H21N5O4.HCl
Formula Weight: 419.86
Physical Form: Powder
Appearance: White
Melting Point: 285°
Beilstein Reference: 4303561
MDL No.: MFCD00058177
Applications: An alpha adrenoceptor antagonist
Vzorec:
C₁₉H₂₁N₅O₄ MW: 419,87 g/mol Bod tání: 285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058177 CAS číslo: 19237-84-4 EINECS: 242-903-4 Merck Index: 14,7717 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...