Tisk…

Acetyl-L-carnitine hydrochloride

Dodavatel: Thermo Scientific
Varování

Synonyms: (R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride, (R)-2-Acetoxy-3-carboxy-N,N,N-trimethylpropan-1-aminium chloride, Acetyl-L-carnitine HCl

J61536.06 J61536.14
J61536.06EA 2650 CZK
J61536.06 J61536.14
Acetyl-L-carnitine hydrochloride
Acetyl-L-carnitine hydrochloride

CAS No.: 5080-50-2 Molecular Formula: C9H17NO4.HCl Formula Weight: 239.70 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Sensitivity: Hygroscopic Appearance: White Beilstein Reference: 4340103 MDL No.: MFCD00082230


Applications: Weak cholinergic agonist

Vzorec: C₉H₁₇NO₄
MW: 239,70 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00082230
CAS číslo: 5080-50-2
Merck Index: 14,84

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR