Telmisartan 99%
Dodavatel: Thermo Scientific
Synonyms:
4'-[[4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylic Acid, 4'-((1,7'-Dimethyl-2'-propyl-1H,3'H-[2,5'-bibenzo[d]imidazol]-3'-yl)methyl)-[1,1'-biphenyl]-2-carboxylic acid
J61441.MD
J61441.03
J61441.MDEA
2380
CZK
J61441.MD
J61441.03
Telmisartan 99%
Telmisartan
Purity: 99%
CAS No.: 144701-48-4
Molecular Formula: C33H30N4O2
Formula Weight: 514.62
Physical Form: Powder
Appearance: White
Melting Point: 261-263°
MDL No.: MFCD00918125
Applications: An angiotensin II receptor antagonist
Vzorec:
C₃₃H₃₀N₄O₂ MW: 514,63 g/mol Bod tání: 261…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD00918125 CAS číslo: 144701-48-4 Merck Index: 14,9129 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...