DL-Arginine hydrochloride
Dodavatel: Thermo Fisher Scientific
CAS No.: 32042-43-6
Molecular Formula: C6H14N4O2.HCl
Formula Weight: 210.67
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00064549
Applications: Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism
Vzorec:
C₆H₁₄N₄O₂.HCl Storage Temperature: Ambient |
MDL Number:
MFCD00064549 CAS číslo: 32042-43-6 EINECS: 250-903-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...