Bezafibrate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(4-(2-(4-Chlorobenzamido)ethyl)phenoxy)-2-methylpropanoic acid, 2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]isobutyric Acid
J61412.ME
J61412.03
J61412.06
J61412.MEEA
1040
CZK
J61412.ME
J61412.03
J61412.06
Bezafibrate ≥98%
Bezafibrate
Purity: 98+%
CAS No.: 41859-67-0
Molecular Formula: C19H20ClNO4
Formula Weight: 361.82
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 183-186°
MDL No.: MFCD00078970
Applications: A peroxisome proliferator-activated receptor agonist and hypolipidemic agent
Vzorec:
C₁₉H₂₀ClNO₄ MW: 361,82 g/mol Bod tání: 183…186 °C Storage Temperature: Freezer |
MDL Number:
MFCD00078970 CAS číslo: 41859-67-0 EINECS: 255-567-9 Merck Index: 14,1195 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...