N-Succinimidyl 3-Maleimidobenzoate 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
MBS
Purity: 95%
CAS No.: 58626-38-3
Molecular Formula: C15H10N2O6
Formula Weight: 314.26
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture and Light Sensitive
Appearance: Off-white
Melting Point: 175-177°
Solubility: Soluble in DMF or ethyl acetate.
Beilstein Reference: 1505254
MDL No.: MFCD00005514
Applications: A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates
Vzorec:
C₁₅H₁₀N₂O₆ MW: 314.25 g/mol Bod tání: 175…177 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005514 CAS číslo: 58626-38-3 EINECS: 261-368-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...