Clindamycin hydrochloride monohydrate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
J61409.03
J61409.06
J61409.03EA
6350
CZK
J61409.03
J61409.06
Clindamycin hydrochloride monohydrate ≥97%
Clindamycin hydrochloride monohydrate
CAS No.: 58207-19-5
Molecular Formula: C18H33ClN2O5S.HCl.H2O
Formula Weight: 479.46 (461.44anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water
Beilstein Reference: 4070786
MDL No.: MFCD07793327
Applications: Antibiotic U-28508E, effective against Gram-positive bacteria
Vzorec:
C₁₈H₃₃ClN₂O₅S MW: 461.44 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06411091 CAS číslo: 58207-19-5 EINECS: 244-398-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...