Phenethyl caffeate ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,4-Dihydroxycinnamic Acid Phenethyl Ester, Caffeic Acid Phenethyl Ester, Phenethyl 3-(3,4-dihydroxyphenyl)acrylate, CAPE, Phenethyl 3,4-Dihydroxycinnamate
Purity: 99+%
CAS No.: 104594-70-9
Molecular Formula: C17H16O4
Formula Weight: 284.31
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Sensitivity: Light Sensitive
Appearance: White to yellow
Melting Point: 127-129°
Solubility: Soluble in ethyl acetate at 50mg/ml. Soluble in DMSO and ethanol.
MDL No.: MFCD00866470
Applications: Inhibitor of protein tyrosine kinase, and is cytotoxic to cancer cell lines
Vzorec:
C₁₇H₁₆O₄ MW: 284,31 g/mol Bod tání: 127…129 °C Storage Temperature: Freezer |
MDL Number:
MFCD00866470 CAS číslo: 104594-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...