Tisk…

7-Chlorokynurenic acid ≥99%

Dodavatel: Thermo Scientific

Synonyms: 7-Chloro-4-hydroxyquinoline-2-carboxylic acid, 7-CTKA

J61359.MA J61359.MF
J61359.MAEA 2950 CZK
J61359.MA J61359.MF
7-Chlorokynurenic acid ≥99%
7-Chlorokynurenic acid

Purity: 99+% CAS No.: 18000-24-3 Molecular Formula: C10H6ClNO3 Formula Weight: 223.62 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Solubility: Soluble to 100 mM in DMSO or 1eq. NaOH MDL No.: MFCD00069227


Applications: NMDA receptor antagonist at the glycine site

Vzorec: C₁₀H₆ClNO₃
MW: 223,62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069227
CAS číslo: 18000-24-3
EINECS: 241-913-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR