Tisk…

Carboxy-PTIO potassium salt ≥98%

Dodavatel: Thermo Fisher Scientific

J61316.MA J61316.MF
J61316.MAEA 3950 CZK
J61316.MA J61316.MF
Carboxy-PTIO potassium salt ≥98%
Carboxy-PTIO potassium salt

Purity: 98+% CAS No.: 148819-94-7 Molecular Formula: C14H16KN2O4 Formula Weight: 315.38 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Blue Solubility: Soluble in water. Also soluble in ethanol, methanol, DMSO or DMF. MDL No.: MFCD00216153


Applications: Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase

Vzorec: C₁₄H₁₆KN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00216153
CAS číslo: 148819-94-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR