6-Azauridine
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,2,4-triazine-3,5(2H,4H)-dione, 6-Azuridine
J61309.03
J61309.06
J61309.03EA
3880
CZK
J61309.03
J61309.06
6-Azauridine
6-Azauridine
CAS No.: 54-25-1
Molecular Formula: C8H11N3O6
Formula Weight: 245.19
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 160-161°
MDL No.: MFCD00006472
Vzorec:
C₈H₁₁N₃O₆ MW: 245,19 g/mol Bod tání: 160…161 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006472 CAS číslo: 54-25-1 EINECS: 200-199-6 Merck Index: 14,904 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...