Tisk…

Felodipine

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl Methyl 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, 3-Ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Ethyl Methyl Ester

J61195.MF J61195.MC J61195.MD
J61195.MFEA 4600 CZK
J61195.MF J61195.MC J61195.MD
Felodipine
Felodipine

CAS No.: 72509-76-3 Molecular Formula: C18H19Cl2NO4 Formula Weight: 384.25 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: Pale yellow to yellow Solubility: Soluble in DMSO. Insoluble in water MDL No.: MFCD00868316


Applications: A dihydropyridine calcium channel blocker

Vzorec: C₁₈H₁₉Cl₂NO₄
MW: 384,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00868316
CAS číslo: 72509-76-3
Merck Index: 14,3953

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR