SKF-96365 hydrochloride ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(2-(4-Methoxyphenyl)-2-(3-(4-methoxyphenyl)propoxy)ethyl)-1H-imidazole hydrochloride, 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole hydrochloride, SKF-96365
J60937.MA
J60937.MF
J60937.MAEA
8760
CZK
J60937.MA
J60937.MF
SKF-96365 hydrochloride ≥99%
SKF-96365 hydrochloride
Purity: 99+%
CAS No.: 130495-35-1
Molecular Formula: C22H26N2O3.HCl
Formula Weight: 402.92
Physical Form: Powder
Appearance: White
Solubility: Soluble to 100mM in water
MDL No.: MFCD00236407
Applications: Inhibits receptor-mediated calcium influx
Vzorec:
C₂₂H₂₆N₂O₃·HCl MW: 402,9 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236407 CAS číslo: 130495-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...