Fasudil dihydrochloride ≥99%
Dodavatel: Thermo Scientific
Synonyms:
1-(5-Isoquinolinesulphonyl)homopiperazine dihydrochloride salt, HA-1077 dihydrochloride salt
J60594.MD
J60594.ME
J60594.03
J60594.MDEA
9130
CZK
J60594.MD
J60594.ME
J60594.03
Fasudil dihydrochloride ≥99%
Fasudil dihydrochloride
Purity: 99+%
CAS No.: 203911-27-7
Molecular Formula: C14H17N3O2S.2HCl
Formula Weight: 364.29
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00153805
Applications: A vasodilator agent which inhibits vascular smooth muscle contraction by acting as an intracellular calcium antagonist
Vzorec:
C₁₄H₁₇N₃O₂S·2HCl MW: 364,29 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00153805 CAS číslo: 203911-27-7 Merck Index: 14,3942 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...