Adenosine-2',3'-cyclic monophosphate sodium salt 97%
Dodavatel: Thermo Scientific
J60558.MC
J60558.MD
J60558.MCEA
6280
CZK
J60558.MC
J60558.MD
Adenosine-2',3'-cyclic monophosphate sodium salt 97%
Adenosine-2',3'-cyclic monophosphate sodium salt
Purity: 97%
CAS No.: 37063-35-7
Molecular Formula: C10H11N5NaO6P
Formula Weight: 351.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 241-243°
MDL No.: MFCD00005757
Vzorec:
C₁₀H₁₁N₅NaO₆P MW: 351,19 g/mol Bod tání: 241…243 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005757 CAS číslo: 37063-35-7 EINECS: 253-328-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...