Meclofenamic acid sodium salt ≥99%
Dodavatel: Thermo Fisher Scientific
J60484.03
J60484.06
J60484.03EA
1510
CZK
J60484.03
J60484.06
Meclofenamic acid sodium salt ≥99%
Meclofenamic acid sodium salt
Purity: 99+%
CAS No.: 6385-02-0
Molecular Formula: C14H10Cl2NNaO2
Formula Weight: 318.13
Physical Form: Crystalline solid
Appearance: White
Solubility: Soluble in water at 50mg/ml. Also soluble in DMF, DMSO or ethanol
MDL No.: MFCD00077376
Applications: Inhibits 5-lipoxygenase and cyclooxygenase; NSAID
Vzorec:
C₁₄H₁₀Cl₂NNaO₂ Storage Temperature: Freezer |
MDL Number:
MFCD00077376 CAS číslo: 6385-02-0 EINECS: 228-983-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...