Tioconazole ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(2-((2-Chlorothiophen-3-yl)methoxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole
J60459.03
J60459.06
J60459.03EA
3950
CZK
J60459.03
J60459.06
Tioconazole ≥98%
Tioconazole
Purity: 98+%
CAS No.: 65899-73-2
Molecular Formula: C16H13Cl3N2OS
Formula Weight: 387.71
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00057276
Applications: Potent inhibitor of cytochrome-P450
Vzorec:
C₁₆H₁₃Cl₃N₂OS MW: 387,71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057276 CAS číslo: 65899-73-2 EINECS: 265-973-8 Merck Index: 14,9452 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...