Quinidine sulphate dihydrate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(S)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol sulphate hydrate(2:1:2)
A sodium channel blocker, and to a lesser extent also blocks potassium channels
Soluble in chloroform, water (11 mg/ml at 20°C), alcohol, and ethanol (50 mg/ml).
Applications: A sodium channel blocker, and to a lesser extent also blocks potassium channels
Vzorec:
C₄₀H₄₈N₄O₄.H₂O₄S·2H₂O MW: 746,92 g/mol Bod tání: 212…214 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00149346 CAS číslo: 6591-63-5 EINECS: 200-046-3 Merck Index: 14,8060 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...