Doxycycline hydrochloride
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(4S,4aR,5S,5aR,6R,12aS)-4-(Dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
J60422.06
J60422.14
J60422.06EA
2970
CZK
J60422.06
J60422.14
Doxycycline hydrochloride
Doxycycline hydrochloride
CAS No.: 10592-13-9
Molecular Formula: C22H24N2O8.HCl
Formula Weight: 480.90
Physical Form: Crystalline powder
Appearance: Yellow
Melting Point: 195-201°
Solubility: Soluble in water to 50mg/ml. May require warming
MDL No.: MFCD03427564
Applications: Inhibits angiogenesis and reduces lung metastases by inhibiting matrix metalloproteinases
Vzorec:
C₂₂H₂₄N₂O₁₀·HCl MW: 480,9 g/mol Bod tání: 195…201 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03427564 CAS číslo: 10592-13-9 EINECS: 234-198-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...