Tisk…

Epirubicin hydrochloride

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (8S,10S)-10-(((2R,4S,5R,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride, Epirubicin HCl

J60411.MA J60411.MC
J60411.MAEA 8140 CZK
J60411.MA J60411.MC
Epirubicin hydrochloride
Epirubicin hydrochloride

CAS No.: 56390-09-1 Molecular Formula: C27H29NO11.HCl Formula Weight: 579.98 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Red Melting Point: 185° Solubility: Soluble in DMSO to 100mM, or in ethanol to 10mM MDL No.: MFCD00941448


Applications: Cell-permeable anthracycline anti-tumor antibiotic

Vzorec: C₂₇H₂₉NO₁₁·HCl
Bod tání: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941448
CAS číslo: 56390-09-1
EINECS: 260-145-2
Merck Index: 14,3623

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR