Clofibrate ≥95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450s CYP4A1 and CYP4A3.
- Applications: Peroxisome proliferator-activated receptor-a agonist. Induces apoptosis in hepatoma cells
Purity: 95+%
CAS No.: 637-07-0
Molecular Formula: C12H15ClO3
Formula Weight: 242.70
Storage Temperature: 2°C to 8°C
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.14
Refractive Index: 1.5
Appearance: Clear
Boiling Point: 154-156°
Beilstein Reference: 1913459
UN No.: UN3082
MDL No.: MFCD00000615
A lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL.
Vzorec:
C₁₂H₁₅ClO₃ Bod varu: 154…156 °C Hustota: 1,14 Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000615 CAS číslo: 637-07-0 EINECS: 211-277-4 UN: 3082 ADR: 9,III Merck Index: 14,2377 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...