Palatinose hydrate ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
6-O-alpha-D-Glucopyranosyl-D-fructose hydrate, Isomaltulose hydrate, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(((2R,3S,4S,5R)-3,4,5-trihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)methoxy)tetrahydro-2H-pyran-3,4,5-triol hydrate
Purity: 98+%
CAS No.: 13718-94-0
Molecular Formula: C12H22O11.xH2O
Formula Weight: 342.30
Physical Form: Powder
Appearance: White
Melting Point: 122-124°
Beilstein Reference: 1757215
MDL No.: MFCD00076094
Vzorec:
C₁₂H₂₂O₁₁·xH₂O MW: 342.30 g/mol (anhy) Bod tání: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076094 CAS číslo: 343336-76-5 EINECS: 237-282-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...