Enrofloxacin
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, 1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS No.: 93106-60-6
Molecular Formula: C19H22FN3O3
Formula Weight: 359.39
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 221-226°
Solubility: Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH
Beilstein Reference: 5307824
MDL No.: MFCD00792463
Applications: A plant fungicide and antibacterial agent
Vzorec:
C₁₉H₂₂FN₃O₃ MW: 359,40 g/mol Bod tání: 221…226 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00792463 CAS číslo: 93106-60-6 Merck Index: 14,3592 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...