Tisk…

Naloxone hydrochloride 98%

Dodavatel: Thermo Fisher Scientific
Varování

J60013.03 J60013.MC J60013.ME
J60013.03EA 15210 CZK
J60013.03 J60013.MC J60013.ME
Naloxone hydrochloride 98%
Naloxone hydrochloride

Purity: 98% CAS No.: 357-08-4 Molecular Formula: C19H21NO4.HCl Formula Weight: 363.84 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White Melting Point: 200-205° MDL No.: MFCD00069322


Applications: A specific opiate antagonist. Blocks the action of sigma-agonists at opioid sites

Vzorec: C₁₉H₂₂ClNO₄
Bod tání: 200…205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069322
CAS číslo: 357-08-4
EINECS: 206-611-0
Merck Index: 14,6362

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR