Tisk…

4,4'-Biphenyldiboronic acid bis(pinacol)ester 95%

Dodavatel: Thermo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

H51956.03 H51956.06
H51956.03EA 1940 CZK
H51956.03 H51956.06
4,4'-Biphenyldiboronic acid bis(pinacol)ester 95%
4,4'-Biphenyldiboronic acid bis(pinacol)ester

Purity: 95% CAS No.: 207611-87-8 Molecular Formula: C24H32B2O4 Formula Weight: 406.14 MDL No.: MFCD08669545

Vzorec: C₂₄H₃₂B₂O₄
MW: 406,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669545
CAS číslo: 207611-87-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR