Tisk…
Propanephosphonic acid cyclic anhydride ≥50% (w/w) in acetonitrile
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
H66967.18 H66967.30
H66967.18EA 2140 CZK
H66967.18 H66967.30
Propanephosphonic acid cyclic anhydride ≥50% (w/w) in acetonitrile
Propanephosphonic acid cyclic anhydride
CAS No.: 68957-94-8 Molecular Formula: C9H21O6P3 Formula Weight: 318.19 Physical Form: Liquid Appearance: Colourless to brown Beilstein Reference: 5079255 UN No.: UN2924 MDL No.: MFCD00006583
Vzorec: C₉H₂₁O₆P₃
MW: 318.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006583
CAS číslo: 68957-94-8
EINECS: 422-210-5
UN: 2924
ADR: 3 II