Benzyl viologen dichloride 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1,1'-Dibenzyl-[4,4'-bipyridine]-1,1'-diium chloride, 1,1'-Dibenzyl-4,4'-bipyridinium dichloride
Purity: 97%
CAS No.: 1102-19-8
Molecular Formula: C24H22Cl2N2
Formula Weight: 409.35
Physical Form: Solid
Appearance: White to off-white
Melting Point: 260° dec.
Beilstein Reference: 3811585
MDL No.: MFCD00011991
Vzorec:
C₂₄H₂₂Cl₂N₂ MW: 409,35 g/mol Bod tání: 260 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011991 CAS číslo: 1102-19-8 EINECS: 214-158-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...