Tisk…
N-Boc-D-aspartic acid-1-methyl ester 97%
Dodavatel: Thermo Fisher Scientific
Synonyms: Boc-D-Asp-OMe, (R)-3-(Boc-amino)-4-methoxy-4-oxobutyric acid
H66573.14 H66573.06
H66573.14EA 9410 CZK
H66573.14 H66573.06
N-Boc-D-aspartic acid-1-methyl ester 97%
N-Boc-D-aspartic acid-1-methyl ester
Purity: 97% CAS No.: 137130-65-5 Molecular Formula: C10H17NO6 Formula Weight: 247.25 Physical Form: Powder Appearance: White
Vzorec: C₁₀H₁₇NO₆
MW: 247.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094776
CAS číslo: 137130-65-5