1-tert-Butylpiperazine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H66497.03
H66497.06
H66497.03EA
1910
CZK
H66497.03
H66497.06
1-tert-Butylpiperazine 97%
1-tert-Butylpiperazine
Purity: 97%
CAS No.: 38216-72-7
Molecular Formula: C8H18N2
Formula Weight: 142.24
Physical Form: Low melting solid
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Colourless to yellow
Melting Point: 32-35°
Boiling Point: 85-87°/22mm
UN No.: UN3259
MDL No.: MFCD00049359
Vzorec:
C₈H₁₈N₂ MW: 142,24 g/mol Bod varu: 85…87 °C (22 mmHg) Bod tání: 32…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049359 CAS číslo: 38216-72-7 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...