2-Chloro-6-(trichloromethyl)pyridine 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Nitrapyrin
H66418.03
H66418.06
H66418.03EA
1480
CZK
H66418.03
H66418.06
2-Chloro-6-(trichloromethyl)pyridine 98%
2-Chloro-6-(trichloromethyl)pyridine
Purity: 98%
CAS No.: 1929-82-4
Molecular Formula: C6H3Cl4N
Formula Weight: 230.90
Flash Point: 100°(212°F)
Physical Form: Powder
Appearance: White
Melting Point: 60-65°
Beilstein Reference: 1618997
UN No.: UN3077
MDL No.: MFCD00072500
Vzorec:
C₆H₃Cl₄N MW: 230,897 g/mol Bod tání: 60…65 °C Bod vzplanutí: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00072500 CAS číslo: 1929-82-4 EINECS: 217-682-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...