2-Amino-1-cyclopentene-1-carbonitrile 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Aminocyclopent-1-enecarbonitrile
H66219.14
H66219.22
H66219.14EA
7550
CZK
H66219.14
H66219.22
2-Amino-1-cyclopentene-1-carbonitrile 98%
2-Amino-1-cyclopentene-1-carbonitrile
Purity: 98%
CAS No.: 2941-23-3
Molecular Formula: C6H8N2
Formula Weight: 108.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
Melting Point: 147-148°
Beilstein Reference: 742061
UN No.: UN3439
MDL No.: MFCD00517551
Vzorec:
C₆H₈N₂ MW: 108,144 g/mol Bod tání: 147…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00517551 CAS číslo: 2941-23-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...