Tisk…

Diethyl-2-thienylmethylphosphonate 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Diethyl (thiophen-2-ylmethyl)phosphonate

H65232.03 H65232.06
H65232.03EA 3510 CZK
H65232.03 H65232.06
Diethyl-2-thienylmethylphosphonate 95%
Diethyl-2-thienylmethylphosphonate

Purity: 95% CAS No.: 2026-42-8 Molecular Formula: C9H15O3PS Formula Weight: 234.25 Physical Form: Liquid Refractive Index: 1.5200 Appearance: Pale yellow Beilstein Reference: 1683411 MDL No.: MFCD00128457

Vzorec: C₉H₁₅O₃PS
Storage Temperature: Ambient
MDL Number: MFCD00128457
CAS číslo: 2026-42-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR