Z-Gln-Gly-OH 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
N-Benzyloxycarbonyl-L-glutaminylglycine, (S)-2-(5-Amino-2-(((benzyloxy)carbonyl)amino)-5-oxopentanamido)acetic acid
H65088.MD
H65088.03
H65088.06
H65088.MDEA
2150
CZK
H65088.MD
H65088.03
H65088.06
Z-Gln-Gly-OH 98%
Z-Gln-Gly-OH
Purity: 98%
CAS No.: 6610-42-0
Molecular Formula: C15H19N3O6
Formula Weight: 337.33
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 2179068
MDL No.: MFCD00055926
Vzorec:
C₁₅H₁₉N₃O₆ Storage Temperature: Freezer |
MDL Number:
MFCD00055926 CAS číslo: 6610-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...