2-Bromo-3'-chloropropiophenone 98%
Dodavatel: Thermo Scientific
|
Varování
|
H64999.03
H64999.06
H64999.03EA
1350
CZK
H64999.03
H64999.06
2-Bromo-3'-chloropropiophenone 98%
2-Bromo-3'-chloropropiophenone
Purity: 98%
CAS No.: 34911-51-8
Molecular Formula: C9H8BrClO
Formula Weight: 247.52
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.532
Refractive Index: 1.5770
Appearance: Colourless to pale yellow
Beilstein Reference: 2360404
MDL No.: MFCD03701150
Vzorec:
C₉H₈BrClO MW: 247,516 g/mol Hustota: 1,532 Storage Temperature: Refrigerator |
MDL Number:
MFCD03701150 CAS číslo: 34911-51-8 EINECS: 252-282-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...